Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP5K59B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043101.4 | NP_001036566.4 | 8 - 9 | 1606 | 102 | |
NM_001259542.2 | NP_001246471.1 | 8 - 9 | 1594 | 102 | ||
NM_001259543.2 | NP_001246472.1 | 8 - 9 | 1594 | 102 | ||
NM_001382080.1 | NP_001369113.1 | 8 - 9 | 1594 | 102 | ||
NM_137885.4 | NP_611729.2 | 8 - 9 | 1594 | 102 | ||
NM_166567.4 | NP_726262.2 | 8 - 9 | 1594 | 102 | ||
GenBank mRNA | BT044218.1 | - | 8 - 9 | 1573 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |