Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078796.3 | NP_523520.2 | 5 - 6 | 927 | 78 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |