Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032072.2 | NP_001027243.2 | 11 - 12 | 1909 | 57 | |
NM_078817.4 | NP_523541.2 | 13 - 14 | 2605 | 57 | ||
GenBank mRNA | BT010016.1 | - | 13 - 14 | 2604 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |