Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014482.3 | NP_001014482.2 | 3 - 4 | 960 | 66 | |
NM_001259102.2 | NP_001246031.1 | 3 - 4 | 960 | 66 | ||
NM_001299002.1 | NP_001285931.1 | 3 - 4 | 960 | 66 | ||
NM_078841.5 | NP_523565.2 | 3 - 4 | 960 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |