Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: beat-Ib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169516.2 | NP_001162987.1 | 6 - 7 | 1930 | 121 | |
NM_078855.4 | NP_523579.1 | 6 - 7 | 1922 | 121 | ||
GenBank mRNA | AF325711.1 | - | 6 - 7 | 1921 | 121 | |
BT050436.1 | - | 6 - 7 | 1928 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |