Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhc36C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382179.1 | NP_001369131.1 | 8 - 9 | 4378 | 70 | |
NM_078867.4 | NP_523591.2 | 8 - 9 | 4378 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |