Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp42Ef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078908.3 | NP_523632.2 | 4 - 5 | 968 | 56 | |
GenBank mRNA | AY070898.1 | - | 4 - 5 | 966 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |