Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp42Eg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078909.4 | NP_523633.1 | 4 - 5 | 760 | 94 | |
GenBank mRNA | AF274017.1 | - | 4 - 5 | 742 | 94 | |
AY113403.1 | - | 4 - 5 | 758 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |