Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp42Ei | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273827.1 | NP_001260756.1 | 2 - 3 | 456 | 74 | |
NM_078911.4 | NP_523635.2 | 2 - 3 | 456 | 74 | ||
GenBank mRNA | AY070990.1 | - | 2 - 3 | 454 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |