Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp42El | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078914.3 | NP_523638.1 | 2 - 3 | 397 | 62 | |
GenBank mRNA | AF274019.1 | - | 2 - 3 | 351 | 62 | |
AY051496.1 | - | 2 - 3 | 351 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |