Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr43a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043066.1 | NP_001036531.1 | 6 - 7 | 945 | 65 | |
NM_001299229.1 | NP_001286158.1 | 6 - 7 | 957 | 65 | ||
NM_078926.3 | NP_523650.2 | 7 - 8 | 879 | 65 | ||
GenBank mRNA | BT023089.1 | - | 7 - 8 | 879 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Drosophila melanogaster |