Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259305.1 | NP_001246234.1 | 4 - 5 | 625 | 82 | |
NM_078952.3 | NP_523676.1 | 4 - 5 | 625 | 82 | ||
GenBank mRNA | AF145673.1 | - | 4 - 5 | 625 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |