Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or47a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078965.3 | NP_523689.1 | 1 - 2 | 316 | 80 | |
GenBank mRNA | AF156880.1 | - | 1 - 2 | 324 | 80 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Recommended Assay Drosophila melanogaster |