Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or47b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078966.3 | NP_523690.3 | 4 - 5 | 1034 | 75 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |