Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144145.4 | NP_001137617.2 | 5 - 6 | 3609 | 72 | |
NM_001201956.2 | NP_001188885.2 | 5 - 6 | 3609 | 72 | ||
NM_001299264.1 | NP_001286193.1 | 5 - 6 | 3609 | 72 | ||
NM_136533.2 | NP_610377.1 | 4 - 5 | 2522 | 72 | ||
GenBank mRNA | AY122074.1 | - | 4 - 5 | 2522 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |