Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cirl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144148.4 | NP_001137620.1 | 1 - 2 | 350 | 71 | |
NM_001169619.4 | NP_001163090.1 | 1 - 2 | 350 | 71 | ||
NM_001259275.2 | NP_001246204.1 | 1 - 2 | 350 | 71 | ||
NM_001259276.2 | NP_001246205.1 | 3 - 4 | 1215 | 71 | ||
NM_001273878.1 | NP_001260807.1 | 4 - 5 | 1041 | 71 | ||
NM_001273879.2 | NP_001260808.1 | 4 - 5 | 1045 | 71 | ||
NM_001273880.2 | NP_001260809.1 | 4 - 5 | 906 | 71 | ||
NM_136553.5 | NP_610397.4 | 4 - 5 | 981 | 71 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |