Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rme-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299307.1 | NP_001286236.1 | 16 - 17 | 5510 | 86 | |
NM_136623.2 | NP_610467.1 | 16 - 17 | 5513 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |