Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4p3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136629.3 | NP_610473.1 | 4 - 5 | 621 | 117 | |
GenBank mRNA | AY075201.1 | - | 4 - 5 | 600 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |