Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144152.2 | NP_001137624.1 | 8 - 9 | 2103 | 119 | |
NM_001144153.2 | NP_001137625.1 | 8 - 9 | 2103 | 119 | ||
NM_001144154.2 | NP_001137626.1 | 6 - 7 | 1382 | 119 | ||
NM_001169627.2 | NP_001163098.1 | 8 - 9 | 2103 | 119 | ||
NM_001299320.1 | NP_001286249.1 | 6 - 7 | 714 | 119 | ||
NM_001299321.1 | NP_001286250.1 | 8 - 9 | 2103 | 119 | ||
NM_136647.5 | NP_610491.4 | 6 - 7 | 1382 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |