Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144152.2 | NP_001137624.1 | 12 - 13 | 5071 | 71 | |
NM_001144153.2 | NP_001137625.1 | 12 - 13 | 5071 | 71 | ||
NM_001144154.2 | NP_001137626.1 | 10 - 11 | 4350 | 71 | ||
NM_001169627.2 | NP_001163098.1 | 12 - 13 | 5068 | 71 | ||
NM_001299320.1 | NP_001286249.1 | 10 - 11 | 3679 | 71 | ||
NM_001299321.1 | NP_001286250.1 | 12 - 13 | 5068 | 71 | ||
NM_136647.5 | NP_610491.4 | 10 - 11 | 4350 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |