Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136726.3 | NP_610570.1 | 1 - 2 | 101 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |