Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273949.1 | NP_001260878.1 | 1 | 636 | 69 | |
NM_136786.3 | NP_610630.1 | 1 | 636 | 69 | ||
GenBank mRNA | AY071329.1 | - | 1 | 634 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |