Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpr47Eb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136811.4 | NP_610655.1 | 1 | 773 | 95 | |
GenBank mRNA | AY070611.1 | - | 1 | 771 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |