Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136878.3 | NP_610722.2 | 3 - 4 | 757 | 67 | |
NM_136879.3 | NP_610723.2 | 3 - 4 | 752 | 67 | ||
GenBank mRNA | BT031295.1 | - | 3 - 4 | 753 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |