Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpr49Ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136925.3 | NP_610769.1 | 2 - 3 | 547 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |