Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dgt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299420.1 | NP_001286349.1 | 2 - 3 | 445 | 72 | |
NM_136941.3 | NP_610785.1 | 3 - 4 | 532 | 72 | ||
GenBank mRNA | AY052018.1 | - | 2 - 3 | 421 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |