Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dgt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299420.1 | NP_001286349.1 | 4 - 5 | 1870 | 81 | |
NM_136941.3 | NP_610785.1 | 5 - 6 | 1957 | 81 | ||
GenBank mRNA | AY052018.1 | - | 4 - 5 | 1846 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |