Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169803.2 | NP_001163274.1 | 14 - 15 | 2262 | 75 | |
NM_001169804.2 | NP_001163275.2 | 10 - 11 | 2422 | 75 | ||
NM_001169805.2 | NP_001163276.1 | 13 - 14 | 2089 | 75 | ||
NM_001169807.2 | NP_001163278.2 | 13 - 14 | 2375 | 75 | ||
NM_137970.5 | NP_611814.3 | 14 - 15 | 2093 | 75 | ||
NM_166616.3 | NP_726350.1 | 13 - 14 | 2375 | 75 | ||
NM_166617.2 | NP_726351.1 | 11 - 12 | 1509 | 75 | ||
NM_166618.3 | NP_726352.1 | 12 - 13 | 1858 | 75 | ||
NM_166619.2 | NP_726353.1 | 1 - 2 | 382 | 75 | ||
GenBank mRNA | AY060327.1 | - | 1 - 2 | 382 | 75 | |
BT011336.1 | - | 13 - 14 | 2339 | 75 | ||
BT021326.1 | - | 14 - 15 | 2089 | 75 | ||
BT133111.1 | - | 12 - 13 | 1858 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |