Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169809.2 | NP_001163280.1 | 4 - 5 | 1098 | 60 | |
NM_001169810.2 | NP_001163281.1 | 4 - 5 | 869 | 60 | ||
NM_058051.4 | NP_477399.1 | 4 - 5 | 1095 | 60 | ||
NM_058052.4 | NP_477400.1 | 4 - 5 | 866 | 60 | ||
NM_134318.3 | NP_599145.1 | 3 - 4 | 1898 | 60 | ||
NM_166628.3 | NP_726373.1 | 4 - 5 | 1095 | 60 | ||
GenBank mRNA | AY089497.1 | - | 4 - 5 | 854 | 60 | |
BT022083.1 | - | 4 - 5 | 1118 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |