Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alpha-Catr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143816.3 | NP_652073.2 | 4 - 5 | 2392 | 111 | |
NM_166641.2 | NP_726389.4 | 4 - 5 | 2392 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |