Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057877.4 | NP_477225.1 | 3 - 4 | 492 | 57 | |
NM_166644.2 | NP_726394.1 | 3 - 4 | 492 | 57 | ||
GenBank mRNA | AF006663.1 | - | 3 - 4 | 440 | 57 | |
AY069103.1 | - | 3 - 4 | 458 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |