Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sei | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299885.1 | NP_001286814.1 | 4 - 5 | 1641 | 93 | |
NM_057365.4 | NP_476713.1 | 3 - 4 | 1160 | 93 | ||
NM_166653.2 | NP_726408.1 | 3 - 4 | 1196 | 93 | ||
GenBank mRNA | AY058350.1 | - | 3 - 4 | 1146 | 93 | |
U36925.1 | - | 3 - 4 | 1131 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |