Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tamo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299898.1 | NP_001286827.1 | 4 - 5 | 2188 | 67 | |
NM_138045.2 | NP_611889.1 | 4 - 5 | 1878 | 67 | ||
NM_166664.1 | NP_726426.1 | 4 - 5 | 2188 | 67 | ||
GenBank mRNA | BT001486.1 | - | 4 - 5 | 2171 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |