Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144281.2 | NP_001137753.1 | 5 - 6 | 1271 | 104 | |
NM_001144282.2 | NP_001137754.1 | 5 - 6 | 1271 | 104 | ||
NM_001299914.1 | NP_001286843.1 | 5 - 6 | 1271 | 104 | ||
NM_166671.2 | NP_726444.2 | 5 - 6 | 1271 | 104 | ||
GenBank mRNA | BT029279.1 | - | 5 - 6 | 1173 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |