Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pof | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259569.2 | NP_001246498.1 | 3 - 4 | 1275 | 99 | |
NM_079127.3 | NP_523851.1 | 3 - 4 | 1279 | 99 | ||
NM_166682.3 | NP_726472.1 | 2 - 3 | 1328 | 99 | ||
GenBank mRNA | AJ295236.1 | - | 2 - 3 | 1256 | 99 | |
AY052060.1 | - | 2 - 3 | 1235 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |