Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_166683.2 | NP_726473.2 | 5 - 6 | 2092 | 59 | |
GenBank mRNA | AY118405.1 | - | 5 - 6 | 2088 | 59 | |
BT021303.1 | - | 5 - 6 | 2088 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |