Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NaCP60E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202078.3 | NP_001189007.2 | 17 - 18 | 6291 | 59 | |
NM_001259578.1 | NP_001246507.1 | 14 - 15 | 5392 | 59 | ||
NM_001259579.3 | NP_001246508.1 | 17 - 18 | 6554 | 59 | ||
NM_001259580.2 | NP_001246509.1 | 18 - 19 | 6306 | 59 | ||
NM_001259581.2 | NP_001246510.1 | 17 - 18 | 6554 | 59 | ||
NM_001274243.2 | NP_001261172.1 | 17 - 18 | 6291 | 59 | ||
NM_001274244.2 | NP_001261173.1 | 16 - 17 | 7496 | 59 | ||
NM_001274245.2 | NP_001261174.1 | 16 - 17 | 6348 | 59 | ||
NM_001274246.2 | NP_001261175.1 | 17 - 18 | 6291 | 59 | ||
NM_166696.5 | NP_726495.3 | 17 - 18 | 6554 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |