Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NaCP60E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202078.3 | NP_001189007.2 | 20 - 21 | 6915 | 142 | |
NM_001259578.1 | NP_001246507.1 | 17 - 18 | 6016 | 142 | ||
NM_001259579.3 | NP_001246508.1 | 20 - 21 | 7178 | 142 | ||
NM_001259580.2 | NP_001246509.1 | 21 - 22 | 6930 | 142 | ||
NM_001259581.2 | NP_001246510.1 | 20 - 21 | 7178 | 142 | ||
NM_001274243.2 | NP_001261172.1 | 20 - 21 | 6915 | 142 | ||
NM_001274244.2 | NP_001261173.1 | 19 - 20 | 8120 | 142 | ||
NM_001274246.2 | NP_001261175.1 | 20 - 21 | 6915 | 142 | ||
NM_166696.5 | NP_726495.3 | 20 - 21 | 7178 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |