Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asator | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272126.2 | NP_001259055.1 | 5 - 6 | 1697 | 70 | |
NM_001272127.1 | NP_001259056.1 | 5 - 6 | 2284 | 70 | ||
NM_001272128.1 | NP_001259057.1 | 5 - 6 | 2284 | 70 | ||
NM_001272129.1 | NP_001259058.1 | 5 - 6 | 1763 | 70 | ||
NM_001297785.1 | NP_001284714.1 | 5 - 6 | 1607 | 70 | ||
NM_143667.3 | NP_651924.2 | 5 - 6 | 1620 | 70 | ||
NM_166765.3 | NP_726576.2 | 5 - 6 | 1620 | 70 | ||
NM_166766.3 | NP_726577.2 | 5 - 6 | 1763 | 70 | ||
GenBank mRNA | BT133187.1 | - | 5 - 6 | 1586 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |