Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014691.3 | NP_001014691.2 | 2 - 3 | 788 | 65 | |
NM_166780.2 | NP_726596.1 | 2 - 3 | 1032 | 65 | ||
NM_166781.2 | NP_726597.1 | 2 - 3 | 788 | 65 | ||
NM_166784.2 | NP_726600.1 | 2 - 3 | 671 | 65 | ||
NM_166785.2 | NP_726601.1 | 2 - 3 | 714 | 65 | ||
GenBank mRNA | AY075429.1 | - | 2 - 3 | 986 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |