Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014691.3 | NP_001014691.2 | 5 - 6 | 2922 | 103 | |
NM_166780.2 | NP_726596.1 | 5 - 6 | 3166 | 103 | ||
NM_166781.2 | NP_726597.1 | 5 - 6 | 2922 | 103 | ||
NM_166784.2 | NP_726600.1 | 5 - 6 | 2805 | 103 | ||
NM_166785.2 | NP_726601.1 | 5 - 6 | 2848 | 103 | ||
GenBank mRNA | AY075429.1 | - | 5 - 6 | 3120 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |