Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PlexA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258499.2 | NP_001245428.1 | 2 - 3 | 1767 | 98 | |
NM_079898.3 | NP_524637.2 | 2 - 3 | 1574 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |