Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trol | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031862.4 | NP_001027033.3 | 11 - 12 | 2130 | 64 | |
NM_001031863.5 | NP_001027034.2 | 6 - 7 | 1022 | 64 | ||
NM_001031865.4 | NP_001027036.3 | 11 - 12 | 2130 | 64 | ||
NM_001031867.4 | NP_001027038.3 | 11 - 12 | 2130 | 64 | ||
NM_001169177.4 | NP_001162648.3 | 10 - 11 | 2010 | 64 | ||
NM_001258562.3 | NP_001245491.2 | 11 - 12 | 2130 | 64 | ||
NM_001258564.3 | NP_001245493.2 | 10 - 11 | 2010 | 64 | ||
NM_001258566.4 | NP_001245495.2 | 7 - 8 | 895 | 64 | ||
NM_001258568.3 | NP_001245497.2 | 10 - 11 | 2010 | 64 | ||
NM_001258570.3 | NP_001245499.2 | 10 - 11 | 2010 | 64 | ||
NM_001258571.3 | NP_001245500.2 | 10 - 11 | 2010 | 64 | ||
NM_001297896.2 | NP_001284825.1 | 10 - 11 | 2010 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |