Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kirre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258576.1 | NP_001245505.1 | 2 - 3 | 1821 | 93 | |
NM_001258577.1 | NP_001245506.1 | 3 - 4 | 1845 | 93 | ||
NM_001258578.1 | NP_001245507.1 | 7 - 8 | 2156 | 93 | ||
NM_001258579.3 | NP_001245508.1 | 7 - 8 | 2026 | 93 | ||
NM_001272267.1 | NP_001259196.1 | 7 - 8 | 2743 | 93 | ||
NM_080320.4 | NP_525059.2 | 9 - 10 | 3223 | 93 | ||
NM_166958.2 | NP_726844.1 | 2 - 3 | 1216 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |