Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103403.1 | NP_001096873.1 | 2 - 3 | 306 | 80 | |
NM_001297914.1 | NP_001284843.1 | 1 - 2 | 37 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |