Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdr50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079016.3 | NP_523740.3 | 5 - 6 | 2993 | 65 | |
GenBank mRNA | BT021362.1 | - | 5 - 6 | 2993 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |