Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RpI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079019.3 | NP_523743.1 | 3 - 4 | 2171 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |