Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079028.3 | NP_523752.1 | 1 | 207 | 91 | |
GenBank mRNA | AY075536.1 | - | 1 | 206 | 91 | |
X91060.1 | - | 192 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster Inventoried |