Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079052.4 | NP_523776.1 | 2 - 3 | 551 | 60 | |
GenBank mRNA | AY051984.1 | - | 2 - 3 | 504 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |