Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smurf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274132.1 | NP_001261061.1 | 6 - 7 | 3639 | 69 | |
NM_079055.3 | NP_523779.1 | 6 - 7 | 3639 | 69 | ||
GenBank mRNA | AF216521.1 | - | 6 - 7 | 3616 | 69 | |
AF416571.1 | - | 6 - 7 | 3616 | 69 | ||
AF464851.1 | - | 5 - 6 | 2735 | 69 | ||
BT021410.1 | - | 6 - 7 | 3616 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |